Learn More
BIOHIPPO Human TrkA/NTRK1 Microsample
SDP
Catalog No. 5000315715
Encompass
Human TrkA/NTRK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TrkA/NTRK1 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TrkA/NTRK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TrkA/NTRK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TrkA/NTRK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15715 Supplier BIOHIPPO Supplier No. BHE15210528
Human TrkA/NTRK1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TrkA/NTRK1 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TrkA/NTRK1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TrkA/NTRK1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TrkA/NTRK1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color