Learn More
ACROBiosystems Human TROP-2 25ug
Catalog No. 502534020
Encompass_Preferred
Biotinylated Human TROP-2 Protein FcAvitag (TR2-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 274 (Accession # P09758-1).
Catalog No. 50-253-4020 Supplier ACROBiosystems Supplier No. TR2H82F425UG
Biotinylated Human TROP-2 Protein FcAvitag (TR2-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 274 (Accession # P09758-1).