Learn More
ACROBiosystems This product is intended for immunocapture, biopanning and flow cytometry.
Catalog No. 502231031
Encompass_Preferred
The Human TROP-2-coupled Magnetic Beads is produced by coupling biotinylated TROP-2 heterodimer to streptavidin-conjugated magnetic beads. Because Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L, the biotinylated protein can bind to the SA beads irreversibly. The pre-coupled beads are ready to use for capturing anti-TROP-2 antibody from your sample with high specificity.
This item is not returnable. View return policy
Catalog No. 50-223-1031 Supplier ACROBiosystems Supplier No. MBSK0282MG
The Human TROP-2-coupled Magnetic Beads is produced by coupling biotinylated TROP-2 heterodimer to streptavidin-conjugated magnetic beads. Because Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L, the biotinylated protein can bind to the SA beads irreversibly. The pre-coupled beads are ready to use for capturing anti-TROP-2 antibody from your sample with high specificity.