Learn More
Sino Biological Human TROP2 / TACSTD2 Protein (His & Fc Tag) 1mg
Supplier:  Sino Biological 10428H03H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-7148