Learn More
Biomatik Corporation Human Troponin T Type 2, Cardiac (TNNT2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12211
Encompass_Preferred
Human Troponin T Type 2, Cardiac (TNNT2) Mini sample ELISA Kit for detecting TNNT2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P45379. Gene ID: 7139. Target Synonyms: CMH2; cTnT; TnTc; Cardiomyopathy,Hypertrophic 2; Cardiac muscle troponin T. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8005