Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Trypsin Pan/PRSS1-2-3 ELISA Kit

Catalog No. EH470RBX10
Encompass_Preferred
Change view
Click to view available options
10 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH470RBX10 10 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH470RBX10 Supplier Invitrogen™ Supplier No. EH470RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human Trypsin Pan/PRSS1-2-3 quantitates human Trypsin Pan/PRSS1-2-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Trypsin Pan/PRSS1-2-3.

Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07477, P07478, P35030
Assay Range 30.72-7500 pg/mL
Assay Sensitivity 30 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Alpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsinogen; beta-trypsin; Brain trypsinogen; cationic trypsinogen; digestive zymogen; mesotrypsin; Mesotrypsinogen; MTG; nonfunctional trypsin 1; pancreatic trypsinogen III; protease serine 2 preproprotein; protease, serine 1; protease, serine 2; protease, serine 3; protease, serine, 2 (trypsin 2); protease, serine, 4 (trypsin 4, brain); PRSS1; PRSS2; PRSS3; PRSS4; Serine protease 1; Serine protease 2; Serine protease 3; Serine protease 4; T9; TCR V beta 4.1; TRP1; TRY1; TRY2; TRY3; TRY4; TRY8; TRYP1; TRYP2; Trypsin I; Trypsin II; trypsin III; Trypsin IV; trypsin-1; Trypsin-2; trypsin-3; trypsinogen 1; trypsinogen 2; trypsinogen 4; trypsinogen 5; trypsinogen A; trypsinogen IVAlpha-trypsin chain 1; Alpha-trypsin chain 2; Anionic trypsinogen; beta-trypsin; Brain trypsinogen; cationic trypsinogen; digestive zymogen; mesotrypsin; Mesotrypsinogen; MTG; nonfunctional trypsin 1; pancreatic trypsinogen III; protease serine 2 preproprotein; protease, serine 1; protease, serine 2; protease, serine 3; protease, serine, 2 (trypsin 2); protease, serine, 4 (trypsin 4, brain); PRSS1; PRSS2; PRSS3; PRSS4; Serine protease 1; Serine protease 2; Serine protease 3; Serine protease 4; T9; TCR V beta 4.1; TRP1; TRY1; TRY2; TRY3; TRY4; TRY8; TRYP1; TRYP2; Trypsin I; Trypsin II; trypsin III; Trypsin IV; trypsin-1; Trypsin-2; trypsin-3; trypsinogen 1; trypsinogen 2; trypsinogen 4; trypsinogen 5; trypsinogen A; trypsinogen IV;
Gene ID (Entrez) 5644, 5645, 5646
Gene Symbol PRSS1, PRSS2, PRSS3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Trypsin Pan
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.