Learn More
Biomatik Corporation Human Trypsin (TRY) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10967
Encompass_Preferred
Human Trypsin (TRY) High-sensitive ELISA Kit for detecting TRY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00761. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7671