Learn More
Biomatik Corporation Human Tryptase (TPS) CLIA Kit
Supplier:  Biomatik Corporation EKU10042
Encompass_Preferred
Human Tryptase (TPS) CLIA Kit for detecting TPS in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q15661-1. Gene ID: 7177. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7686