Learn More
BIOHIPPO Human TSH Microsample
SDP
Catalog No. 5000313997
Encompass
Human TSH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSH in Serum It provides a detection range of 0 32-20 uIU/mL with a sensitivity of 0 121 uIU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TSH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TSH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TSH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13997 Supplier BIOHIPPO Supplier No. BHE15212307
Human TSH Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSH in Serum It provides a detection range of 0 32-20 uIU/mL with a sensitivity of 0 121 uIU/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TSH Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TSH Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TSH biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color