Learn More
BIOHIPPO Human TSHR Microsample
SDP
Catalog No. 5000313872
Encompass
Human TSHR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSHR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TSHR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TSHR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TSHR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13872 Supplier BIOHIPPO Supplier No. BHE15210569
Human TSHR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TSHR in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 11 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TSHR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TSHR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TSHR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color