Learn More
ACROBiosystems Human TSLP 100ug
Catalog No. 502534007
Encompass_Preferred
Biotinylated Human TSLP HisAvitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1).
Catalog No. 50-253-4007 Supplier ACROBiosystems Supplier No. TSPH82EB100UG
Biotinylated Human TSLP HisAvitag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1).