Learn More
Sino Biological Recombinant Human TSLP Protein (R127A, R130S, His Tag), HPLC-verified
Supplier:  Sino Biological 16135H08H500
Encompass_Preferred
A DNA sequence encoding the human TSLP (NP_149024.1) (Tyr29-Gln159, with mutations Arg 127 Ala, Arg 130 Ser) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6685