Learn More
Sino Biological Recombinant Human TSLP Protein (R127A, R130S, His Tag), HPLC-verified
Catalog No. 502246686
Encompass_Preferred
A DNA sequence encoding the human TSLP (NP_149024.1) (Tyr29-Gln159, with mutations Arg 127 Ala, Arg 130 Ser) was expressed with a polyhistidine tag at the C-terminus.
This item is not returnable. View return policy
Catalog No. 50-224-6686 Supplier Sino Biological Supplier No. 16135H08H100
A DNA sequence encoding the human TSLP (NP_149024.1) (Tyr29-Gln159, with mutations Arg 127 Ala, Arg 130 Ser) was expressed with a polyhistidine tag at the C-terminus.