Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TSLP Recombinant Protein, PeproTech®

Catalog No. 30062100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30062100UG 100 μg
300-62-10UG 10 μg
300-62-1MG 1 mg
30062250UG 250 μg
300-62-2UG 2 μg
30062500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30062100UG Supplier Gibco™ Supplier No. 30062100UG
  • $1,194.00
    $1,014.65 / Each of 1

    Save $179.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TSLP is a 15.0 kDa protein consisting of 132 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q969D9
For Use With (Application) Control, Western Blot Control, ELISA Standard
Formulation Protein with no preservative
Gene ID (Entrez) 85480
Molecular Weight (g/mol) 15 kDa
Name Human TSLP
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP
Common Name TSLP
Gene Symbol TSLP
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MYDFTNCDFE KIKAAYLSTI SKDLITYMSG TKSTEFNNTV SCSNRPHCLT EIQSLTFNPTAGCASLAKEM FAMKTKAALA IWCPGYSETQ INATQAMKKR RKRKVTTNKC LEQVSQLQGLWRRFNRPLLK QQ
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.