Learn More
Sino Biological Human TSPAN1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13073T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TSPAN1 (rh TSPAN1; Catalog#13073-H07H; O60635; Tyr110-Asn211). TSPAN1 specific IgG was purified by Human TSPAN1 affinity chromatography.
Catalog No. 50-237-3785