Learn More
Sino Biological Human TSPAN7 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 14031MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TALLA-1 / TSPAN7 / CD231 (rh TALLA-1 / TSPAN7 / CD231; Catalog#14031-H08H; AAH18036.1; Arg113-Met213). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1412