Learn More
Sino Biological Recombinant Human TSPAN8 Protein (His Tag)
Supplier:  Sino Biological 15683H07H50
Encompass_Preferred
A DNA sequence encoding the human TSPAN8 (P19075) (Lys110-Asn205) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-224-6962