Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TTC31 (aa 41-129) Control Fragment Recombinant Protein

Catalog No. RP98397
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98397 Supplier Invitrogen™ Supplier No. RP98397
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (28%), Rat (28%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59727. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The TTC31 gene, also known as Tetratricopeptide repeat domain 31, is part of a large protein family characterized by tandem TPR motifs. These motifs are involved in mediating protein-protein interactions and are typically found in proteins implicated in various cellular processes such as cell cycle regulation, transcription, and protein transport. The TPR domain's flexibility allows TTC31 to interact with a wide array of partner proteins, contributing to its role in signaling pathways and cellular homeostasis. While specific studies about TTC31's function are limited, its structural characteristics suggest significant involvement in maintaining cellular integrity and function.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q49AM3
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 64427
Name Human TTC31 (aa 41-129) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias tetratricopeptide repeat domain 31; tetratricopeptide repeat protein 31; TPR repeat protein 31; TTC31
Common Name TTC31
Gene Symbol TTC31
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GEEDIVDFLRRLVESDPQGLHRIHVDGSSGRLQLWHHDYLLGHLDDEGKSTGQSDRGKGAEGLGTYCGLRKSFLYPPQESEPCPQSPSA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.