Learn More
BIOHIPPO Human TTN Microsample
SDP
Catalog No. 5000314255
Encompass
Human TTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TTN in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14255 Supplier BIOHIPPO Supplier No. BHE15212107
Human TTN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TTN in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TTN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TTN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TTN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color