Learn More
BIOHIPPO Human TTR Microsample
SDP
Catalog No. 5000315754
Encompass
Human TTR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TTR in serum plasma tissue homogenates cell lysates cerebrospinal fluid cell culture supernates and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 2 4 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TTR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TTR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TTR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15754 Supplier BIOHIPPO Supplier No. BHE15212637
Human TTR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TTR in serum plasma tissue homogenates cell lysates cerebrospinal fluid cell culture supernates and other biological fluids It provides a detection range of 7 82-500 g/mL with a sensitivity of 2 4 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TTR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TTR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TTR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color