Learn More
Sino Biological Anti-Transthyretin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12091RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Prealbumin / Transthyretin / TTR (rh Prealbumin / Transthyretin / TTR; Catalog#12091-H08H; NP_000362.1; Met 1-Glu 147). Prealbumin / Transthyretin / TTR specific IgG was purified by Human Prealbumin / Transthyretin / TTR affinity chromatography.
Catalog No. 50-241-1608