Learn More
Sino Biological Anti-Transthyretin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12091RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Prealbumin / Transthyretin (rh TTR; Catalog#12091-H08H; NP_000362.1; Met1-Glu147). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0247