Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TTYH2 Synthetic Peptide

Catalog No. PEP1437 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1437 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1437 Supplier Invitrogen™ Supplier No. PEP1437
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 14 amino acid peptide near the amino terminus of human TTYH2. PEP-1437 can be used as a blocking peptide with polyclonal antibody PA5-34395.

TTYH2 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH2 is a Ca2+-activated large conductance chloride (Cl-) channel and may play a role in kidney tumorigenesis. It is also upregulated in colon carcinoma and is thought to play a role in cell proliferation and call aggregation, suggesting that TTYH2 may play a role in regulating both proliferation and metastatic potentials of colorectal cancer. The ubiquitin-protein ligase Nedd4-2 binds to and ubiquitinates TTYH2, thereby regulating its cell surface expression and total cellular levels.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 94015
Species Human
Purification Method purified
Accession Number Q9BSA4
Common Name TTYH2
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1110001A03Rik; C17orf29; hTTY2; mTTY2; Protein tweety homolog 2; RGD1562969; TTYH2; tweety family member 2; tweety homolog 2; tweety homolog 2 (Drosophila)
Gene Symbol TTYH2
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.