Learn More
BIOHIPPO Human TUBb Microsample
SDP
Catalog No. 5000315761
Encompass
Human TUBb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TUBb in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TUBb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TUBb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TUBb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15761 Supplier BIOHIPPO Supplier No. BHE15211347
Human TUBb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TUBb in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TUBb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TUBb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TUBb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color