Learn More
Biomatik Corporation Human Tubulin Alpha 1A (TUBa1A) ELISA Kit
Supplier:  Biomatik Corporation EKU11497
Encompass_Preferred
Human Tubulin Alpha 1A (TUBa1A) ELISA Kit for detecting TUBa1A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q71U36. Gene ID: 7846. Target Synonyms: B-ALPHA-1; TUBA3; Brain-Specific Tubulin,Alpha; Alpha-tubulin 3; Tubulin B-alpha-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8134