Learn More
Biomatik Corporation Human Tubulin Beta 1 (TUBb1) ELISA Kit
Supplier:  Biomatik Corporation EKU11498
Encompass_Preferred
Human Tubulin Beta 1 (TUBb1) ELISA Kit for detecting TUBb1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H4B7. Gene ID: 81027. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6943