Learn More
Biomatik Corporation Human Tuftelin (TUFT) ELISA Kit
Supplier:  Biomatik Corporation EKU11720
Encompass_Preferred
Human Tuftelin (TUFT) ELISA Kit for detecting TUFT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NNX1. Gene ID: 7286. Target Synonyms: TUFT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8092