Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&DSystems™ Human Tumor Biomarker Luminex™ Performance Assay

Catalog No. FCSTM2506
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCSTM2506 6 Plex
FCSTM2508 8 Plex
FCSTM2505 5 Plex
FCSTM2510 10 Plex
FCSTM2509 9 Plex
FCSTM2519 19 Plex
FCSTM2526 26 Plex
FCSTM2516 16 Plex
FCSTM2517 17 Plex
FCSTM2520 20 Plex
FCSTM2518 18 Plex
FCSTM2513 13 Plex
FCSTM2504 4 Plex
FCSTM2503 3 Plex
FCSTM2515 15 Plex
FCSTM2524 24 Plex
FCSTM2511 11 Plex
FCSTM2521 21 Plex
FCSTM2525 25 Plex
FCSTM2507 7 Plex
FCSTM2522 22 Plex
FCSTM2502 2 Plex
FCSTM2512 12 Plex
FCSTM2527 27 Plex
FCSTM2523 23 Plex
FCSTM2514 14 Plex
FCSTM2528 28 Plex
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
This item is not returnable. View return policy
Catalog No. FCSTM2506 Supplier R&DSystems™ Supplier No. FCSTM2506
Add to Cart
Add to Cart
This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ platform

Specifications

Quantity 6 Plex
Species Human
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.