Learn More
R&D Systems™ Human Tumor Biomarker Luminex™ Performance Assay

Description
Magnetic Luminex High Performance Assays are flexible bead-based multiplex assays. They allow up to 50 user-defined target analytes to be simultaneously profiled using cell culture supernates, serum, or plasma samples. Some panels are also validated with human milk, saliva, and/or urine samples.
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay |
| Quantity | 1 Plex |
| Species | Human |
| Binding Target | Multiplex Cytokine Assay Kits |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Oncology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.