Learn More
Sigma Aldrich Fine Chemicals Biosciences Human Tumor Necrosis facto
Catalog No. 502906484
Encompass
The Human TNf-alpha ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human TNf-alpha in serum plasma cell culture supernatants and urine.
Catalog No. 50-290-6484 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB04761KT
The Human TNf-alpha ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human TNf-alpha in serum plasma cell culture supernatants and urine.