Learn More
Biomatik Corporation Human Tumor Necrosis Factor Alpha Induced Protein 6 (TNFaIP6) ELISA Kit
Supplier:  Biomatik Corporation EKU12092
Encompass_Preferred
Human Tumor Necrosis Factor Alpha Induced Protein 6 (TNFaIP6) ELISA Kit for detecting TNFaIP6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98066. Gene ID: 7130. Target Synonyms: TSG6; TNF-Stimulated Gene 6; Hyaluronate-binding protein; Tumor necrosis factor alpha-induced protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7223