Learn More
American Research Products Inc Human Tumor necrosis factor li
Supplier:  American Research Products Inc CSBEL023991HU
Encompass
Human Tumor necrosis factor ligand superfamily member 14 (TNFSF14) ELISA kit from CUSABIO Detection Range 0 156 ng/ml - 10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28191