Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 10C (TNFRSF10C) ELISA Kit
Supplier:  Biomatik Corporation EKU11500
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 10C (TNFRSF10C) ELISA Kit for detecting TNFRSF10C in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14798. Gene ID: 8794. Target Synonyms: CD263; TNFRSF10-C; DCR1; LIT; TRAILR3; TRID; Decoy Receptor 1; Tnf-Related Apoptosis-Inducing Ligand Receptor 3; Trail Receptor Without An Intracellular Domain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7756