Learn More
American Research Products Inc Human Tumor necrosis factor re
Catalog No. 5000327208
Encompass
Human Tumor necrosis factor receptor superfamily member 9 (TNFRSF9) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27208 Supplier American Research Products Inc Supplier No. CSBEL023984HU
Human Tumor necrosis factor receptor superfamily member 9 (TNFRSF9) ELISA kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich