Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 4 (TNFRSF4) ELISA Kit
Supplier:  Biomatik Corporation EKU12178
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 4 (TNFRSF4) ELISA Kit for detecting TNFRSF4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43489. Gene ID: 7293. Target Synonyms: CD134; TNFRSF4; ACT35; OX40; TXGP1L; TAX transcriptionally-activated glycoprotein 1 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8014