Learn More
Biomatik Corporation Human Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) CLIA Kit
Supplier:  Biomatik Corporation EKU08846
Encompass_Preferred
Human Tumor Necrosis Factor Related Apoptosis Inducing Ligand (TRAIL) CLIA Kit for detecting TRAIL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 15.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P50591. Gene ID: 8743. Target Synonyms: CD253; TNFSF10; APO2L; Apo2-L; TL2; TRAIL; Tumor Necrosis Factor Ligand Superfamily Member 10; Apo-2 Ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6108