Learn More
American Research Products Inc Human Tumor necrosis factor so
Catalog No. 5000326308
Encompass
Human Tumor necrosis factor soluble receptor I TNFsR-I ELISA KIT from CUSABIO Detection Range 15 625 pg/mL-1000 pg/mL Sensitivity 1 261 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26308 Supplier American Research Products Inc Supplier No. CSBE04737M
Human Tumor necrosis factor soluble receptor I TNFsR-I ELISA KIT from CUSABIO Detection Range 15 625 pg/mL-1000 pg/mL Sensitivity 1 261 pg/mL Sample serum plasma tissue homogenates Method Sandwich