Learn More
Bioworld HUMAN TUMOR NECROSIS FACTOR
Supplier:  Bioworld 220605051
Encompass_Preferred
HUMAN TUMOR NECROSIS FACTOR-BETA (RECOMBINANT): Recombinant Human Tumor Necrosis Factor-beta (TNF-b) Source: E.Coli Description: Recombinant Human TNF-beta (Lymphotoxin) produced in E.Coli is a single, non-glycosylated, polypeptide c...
Catalog No. 50-253-600