Learn More
Biomatik Corporation Human Tumor Protein P63 (TP63) ELISA Kit
Supplier:  Biomatik Corporation EKU07988
Encompass_Preferred
Human Tumor Protein P63 (TP63) ELISA Kit for detecting TP63 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H3D4. Gene ID: 8626. Target Synonyms: P63; TP73L; CUSP; EEC3; KET; LMS; OFC8; RHS; SHFM4; TP53CP; Tumor Protein p53-Competing Protein; Chronic ulcerative stomatitis protein; Keratinocyte transcription factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8708