Learn More
Biomatik Corporation Human Twinfilin 1 (TWF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07993
Encompass_Preferred
Human Twinfilin 1 (TWF1) ELISA Kit for detecting TWF1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12792. Gene ID: 5756. Target Synonyms: A6; PTK9; Actin-Binding Protein,Homolog 1; Protein Tyrosine Kinase 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8707