Learn More
Sino Biological Human TXN2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 12557T30100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Thioredoxin 2 (rh Thioredoxin 2; Catalog#12557-H07E; Q99757; Thr60-Gly166). Thioredoxin 2 specific IgG was purified by Human Thioredoxin 2 affinity chromatography.
Catalog No. 50-237-2949