Learn More
Sino Biological Human TXNDC15 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 12973H08H1
Encompass_Preferred
A DNA sequence encoding the Human TXNDC15 (NP_078991.3) (Val33-Ala322,V322A) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3668