Learn More
Biomatik Corporation Human TYRO Protein Tyrosine Kinase Binding Protein (TYROBP) ELISA Kit
Supplier:  Biomatik Corporation EKU08368
Encompass_Preferred
Human TYRO Protein Tyrosine Kinase Binding Protein (TYROBP) ELISA Kit for detecting TYROBP in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.053ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43914. Gene ID: 7305. Target Synonyms: DAP12; KARAP; PLOSL; Killer Activating Receptor Associated Protein; DNAX-Activation Protein 12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8674