Learn More
Bioworld HUMAN TYROSINE KINASE ERBB-
Supplier:  Bioworld 220606801
Encompass_Preferred
HUMAN TYROSINE KINASE ERBB-3 (RECOMBINANT): Recombinant Human Tyrosine Kinase receptor ErbB-3 Source: E.Coli Background: ErbB3/Her3 (human epidermal growth factor receptor 3), is a type I membrane glycoprotein that is a member of the ...
Catalog No. 50-253-780