Learn More
Biomatik Corporation Human Tyrosyl DNA Phosphodiesterase 1 (TDP1) ELISA Kit
Supplier:  Biomatik Corporation EKU11505
Encompass_Preferred
Human Tyrosyl DNA Phosphodiesterase 1 (TDP1) ELISA Kit for detecting TDP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NUW8. Gene ID: 55775. Target Synonyms: SCAN1; Tyr-DNA phosphodiesterase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8133