Learn More
American Research Products Inc Human U3 small nucleolar ribon
Catalog No. 5000327513
Encompass
Human U3 small nucleolar ribonucleoprotein protein IMP3 (IMP3) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27513 Supplier American Research Products Inc Supplier No. CSBEL011692HU
Human U3 small nucleolar ribonucleoprotein protein IMP3 (IMP3) ELISA kit from CUSABIO Detection Range 39 pg/mL-2500 pg/mL Sensitivity 9 75 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich