Learn More
Sino Biological Human UBA1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11990MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human UBE1 / UBA1 (rh UBE1 / UBA1; Catalog#11990-H20B; NP_003325.2; Ser2-Arg1058). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2521