Learn More
Ubpbio HUMAN UBE1, 25UG
Catalog No. 501126852
Encompass_Preferred
E1 Activating Enzymes – UbE1 (UBPBio catalog# B1100, B1101) – Purified from sf9 insect cells. UBE1 is an E1 enzyme within the E1, E2, E3 cascade which conjugates Ub to protein substrates. UBE1 activates Ub by catalyzing an ATP – dependent reaction; Ub is conjugated onto the catalytic Cys residue of UBE1 by formation of a thioester bond. The activated Ub is then passed onto a Ub-conjugating enzyme E2; Ub charged E2 binds a Ub ligase E3 that directly binds substrate proteins. Ub is transferred to the HECT domain E3s prior to being conjugated on substrate proteins. For RING domain E3s, Ub is transferred directly from an E2 to substrate proteins.
Catalog No. 50-112-6852 Supplier Ubpbio Supplier No. B1100
E1 Activating Enzymes – UbE1 (UBPBio catalog# B1100, B1101) – Purified from sf9 insect cells. UBE1 is an E1 enzyme within the E1, E2, E3 cascade which conjugates Ub to protein substrates. UBE1 activates Ub by catalyzing an ATP – dependent reaction; Ub is conjugated onto the catalytic Cys residue of UBE1 by formation of a thioester bond. The activated Ub is then passed onto a Ub-conjugating enzyme E2; Ub charged E2 binds a Ub ligase E3 that directly binds substrate proteins. Ub is transferred to the HECT domain E3s prior to being conjugated on substrate proteins. For RING domain E3s, Ub is transferred directly from an E2 to substrate proteins.