Learn More
Sino Biological Anti-Rad6/UBE2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12726T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human UBE2A / HHR6A (rh UBE2A / HHR6A; Catalog#12726-H07E; P49459; Met1-Cys152). UBE2A / HHR6A specific IgG was purified by Human UBE2A / HHR6A affinity chromatography.
Catalog No. 50-240-9921