Learn More
Sino Biological Anti-UBE2W Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12725MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human UBE2W (rh UBE2W; Catalog#12725-H07E; Q96B02-12; Met1-Cys151). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4347